Assay Details
Target Gene Details
Entrez Gene ID: | 58810 |
Gene Name: | aldo-keto reductase family 1, member A1 (aldehyde reductase) |
Gene Aliases: |
2610201A18Rik, Akr1a4 |
Location: |
Chr.4:116636510-116651674 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Akr1a1 | NM_021473.3 | NP_067448.1 | ||
AF225564.1 | AAF67111.1 | |||
AK005376.1 | ||||
AK009462.1 | BAB26303.1 | |||
AK011321.1 | BAB27543.1 | |||
AK011388.1 | BAB27586.1 | |||
AK011667.1 | BAB27767.1 | |||
AK011794.1 | BAB27846.1 | |||
AK011856.1 | BAB27883.1 | |||
AK011906.1 | BAB27907.1 | |||
AK011908.1 | BAB27909.1 | |||
AK011918.1 | BAB27915.1 | |||
AK128906.1 | ||||
AK144198.1 | BAE25762.1 | |||
AK146081.1 | BAE26885.1 | |||
AK146278.1 | BAE27036.1 | |||
AK152086.1 | BAE30936.1 | |||
AK159573.1 | ||||
AK160185.1 | ||||
AK164461.1 | BAE37798.1 | |||
AK168950.1 | BAE40756.1 | |||
AK178751.1 | ||||
AK186333.1 | ||||
AK192030.1 | ||||
AK203039.1 | ||||
AK210646.1 | ||||
AK219186.1 | ||||
AY207463.1 | AAO72145.1 | |||
BC039926.1 | AAH39926.1 | |||
BC046762.1 | AAH46762.1 |
More Information
Set Membership: |
Intragenic Non-exonic |