Assay Details
Target Gene Details
Entrez Gene ID: | 12972 |
Gene Name: | crystallin, zeta |
Gene Aliases: |
Sez9 |
Location: |
Chr.3:154596711-154623182 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cryz | NM_009968.3 | 1 | 342 | NP_034098.1 |
AK133820.1 | 1 | 343 | ||
BB618817.1 | 1 | 343 |
Target Gene Details
Entrez Gene ID: | 209584 |
Gene Name: | tRNA-yW synthesizing protein 3 homolog (S. cerevisiae) |
Gene Aliases: |
5230400J09Rik |
Location: |
Chr.3:154576520-154597098 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tyw3 | NM_001168358.1 | 1 | 47 | NP_001161830.1 |
NM_172474.5 | 1 | 47 | NP_766062.1 | |
AK030344.1 | 1 | 45 | BAC26913.1 | |
AK034820.1 | BAC28842.1 | |||
BC086686.1 | ||||
BC087879.1 | AAH87879.1 | |||
BC141040.1 |
More Information
Set Membership: |
Intragenic Exonic |