Assay Details
Target Gene Details
Entrez Gene ID: | 77739 |
Gene Name: | ADAMTS-like 1 |
Gene Aliases: |
5930437A14Rik, 6720426B09Rik, ADAMTSL-1 |
Location: |
Chr.4:85513234-86428390 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Adamtsl1 | NM_029967.3 | 2 | 275 | NP_084243.3 |
AK020115.1 | 2 | 216 | BAB32000.1 | |
AK045085.1 | 2 | 171 | BAC32213.1 | |
AK081888.1 | ||||
BC062814.1 | 2 | 211 | AAH62814.1 | |
BC139339.1 | 2 | 154 | ||
BC139367.1 | 2 | 154 | ||
CJ051149.1 | 2 | 276 |
More Information
Set Membership: |
Intragenic Exonic |