Assay Details
Target Gene Details
Entrez Gene ID: | 77739 |
Gene Name: | ADAMTS-like 1 |
Gene Aliases: |
5930437A14Rik, 6720426B09Rik, ADAMTSL-1 |
Location: |
Chr.4:85513234-86428390 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Adamtsl1 | NM_029967.3 | 10 | 1367 | NP_084243.3 |
CV555842.1 |
More Information
Set Membership: |
Intragenic Exonic |