Assay Details
Target Gene Details
Entrez Gene ID: | 214191 |
Gene Name: | tetratricopeptide repeat domain 24 |
Gene Aliases: |
A430025D11Rik |
Location: |
Chr.3:88069410-88081987 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ttc24 | NM_172526.3 | NP_766114.1 | ||
AK039884.1 | BAC30472.1 | |||
AK076743.1 | ||||
BC055114.1 | AAH55114.1 | |||
BC147554.1 | ||||
BC147555.1 |