Assay Details
Target Gene Details
Entrez Gene ID: | 242800 |
Gene Name: | tetratricopeptide repeat domain 34 |
Gene Aliases: |
4932418L02, B230396O12Rik |
Location: |
Chr.4:154833443-154867127 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ttc34 | NM_172878.3 | 2 | 309 | NP_766466.3 |
AK030055.1 | 2 | 283 | BAC26761.1 | |
CJ183638.1 | 2 | 312 |