Assay Details
Target Gene Details
Entrez Gene ID: | 56079 |
Gene Name: | astrotactin 2 |
Gene Aliases: |
1d8, Astnl, bM452J22.1 |
Location: |
Chr.4:65380803-66404530 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Astn2 | NM_019514.3 | 13 | 2474 | NP_062387.2 |
NM_207109.2 | 14 | 2630 | NP_996992.1 | |
AF116573.1 | 11 | 1642 | AAF14356.1 | |
AK041160.1 | 15 | 2534 | BAC30843.1 |