Assay Details
Target Gene Details
Entrez Gene ID: | 70465 |
Gene Name: | WD repeat domain 77 |
Gene Aliases: |
2610003I18Rik, 2610312E17Rik, C79984, p44/MEP50 |
Location: |
Chr.3:105959498-105969760 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Wdr77 | NM_027432.3 | NP_081708.1 | ||
AK032897.1 | BAC28076.1 | |||
AK048488.1 | BAC33350.1 | |||
AK050391.1 | BAC34232.1 | |||
AK078856.1 | ||||
AK131608.1 | BAE20717.1 | |||
AK161812.1 | 8 | 1337 | ||
AK168006.1 | ||||
AK168752.1 | ||||
AK169128.1 | BAE40907.1 | |||
BC005755.1 | AAH05755.1 |