Assay Details
Target Gene Details
Entrez Gene ID: | 100317 |
Gene Name: | expressed sequence AU040320 |
Gene Aliases: |
A730047D20Rik, Kiaa0319l |
Location: |
Chr.4:126753555-126869694 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AU040320 | NM_001035525.1 | 7 | 1282 | NP_001030602.1 |
NM_001035526.1 | 7 | 1400 | NP_001030603.1 | |
NM_133886.2 | 7 | 1400 | NP_598647.1 | |
AK031342.1 | 7 | 1344 | ||
AK043006.1 | 7 | 1350 | BAC31432.1 | |
AK049570.1 | 7 | 1283 | BAC33818.1 | |
AK084668.1 | 7 | 1245 | BAC39244.1 | |
AK170261.1 | 7 | 1357 | BAE41669.1 | |
AK197670.1 | ||||
BC028869.1 | 7 | 1370 | AAH28869.1 | |
CB249000.1 | ||||
CB527694.1 | 3 | 202 |