Assay Details
Target Gene Details
Entrez Gene ID: | 100317 |
Gene Name: | expressed sequence AU040320 |
Gene Aliases: |
A730047D20Rik, Kiaa0319l |
Location: |
Chr.4:126753555-126869694 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AU040320 | NM_001035525.1 | 2 | 152 | NP_001030602.1 |
NM_001035526.1 | 2 | 270 | NP_001030603.1 | |
NM_133886.2 | 2 | 270 | NP_598647.1 | |
AK031342.1 | 2 | 214 | ||
AK043006.1 | 2 | 220 | BAC31432.1 | |
AK049570.1 | 2 | 153 | BAC33818.1 | |
AK084668.1 | 2 | 115 | BAC39244.1 | |
AK163033.1 | 2 | 78 | ||
AK170261.1 | 2 | 227 | BAE41669.1 | |
AK172674.1 | 1 | 3348 | ||
AK200946.1 | ||||
AK206544.1 | ||||
AK211401.1 | ||||
BC028869.1 | 2 | 240 | AAH28869.1 | |
BY289654.1 | 2 | 271 | ||
BY308757.1 | 2 | 246 |