Assay Details
Target Gene Details
Entrez Gene ID: | 74183 |
Gene Name: | PPARGC1 and ESRR induced regulator, muscle 1 |
Gene Aliases: |
2310042D19Rik |
Location: |
Chr.4:156203268-156221307 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Perm1 | NM_172417.3 | 3 | 2581 | NP_766005.2 |
AK036660.1 | 3 | 2583 | BAC29522.1 | |
BC117871.1 | 2 | 2455 | ||
BC117872.1 | 2 | 2455 | ||
KF150176.1 | 2 | 2248 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)