Assay Details
Target Gene Details
Entrez Gene ID: | 100317 |
Gene Name: | expressed sequence AU040320 |
Gene Aliases: |
A730047D20Rik, Kiaa0319l |
Location: |
Chr.4:126753555-126869694 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AU040320 | NM_001035525.1 | 10 | 1712 | NP_001030602.1 |
NM_001035526.1 | 10 | 1830 | NP_001030603.1 | |
NM_133886.2 | 10 | 1830 | NP_598647.1 | |
AK031342.1 | 10 | 1774 | ||
AK043006.1 | 10 | 1780 | BAC31432.1 | |
AK049570.1 | 10 | 1713 | BAC33818.1 | |
AK084668.1 | 10 | 1675 | BAC39244.1 | |
AK170261.1 | 10 | 1787 | BAE41669.1 | |
AK179638.1 | 1 | 126 | ||
AK208774.1 | 2 | 272 | ||
AK215400.1 | 1 | 126 | ||
BC022154.1 | 1 | 242 | AAH22154.1 | |
BC028869.1 | 10 | 1800 | AAH28869.1 | |
CA540237.1 | 1 | 136 | ||
CB527694.1 |