Assay Details
Target Gene Details
Entrez Gene ID: | 69893 |
Gene Name: | cytochrome c oxidase assembly factor 7 |
Gene Aliases: |
2010305A19Rik, AI451324, D4Ertd796e, Selrc1 |
Location: |
Chr.4:108328152-108340718 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Coa7 | NM_027250.4 | NP_081526.1 | ||
AK008518.1 | ||||
AK044999.1 | BAC32178.1 | |||
AK081388.1 | BAC38209.1 | |||
AK168344.1 | BAE40281.1 | |||
BC012391.1 | AAH12391.1 |