Assay Details
Target Gene Details
Entrez Gene ID: | 11303 |
Gene Name: | ATP-binding cassette, sub-family A (ABC1), member 1 |
Gene Aliases: |
ABC-1, Abc1 |
Location: |
Chr.4:53030789-53159988 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 46 - Exon 46 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Abca1 | NM_013454.3 | NP_038482.3 | ||
CD351490.1 | ||||
CN702062.1 | ||||
X75926.1 | CAA53530.1 |
More Information
Set Membership: |
Intragenic Non-exonic |