Assay Details
Target Gene Details
Entrez Gene ID: | 22253 |
Gene Name: | unc-5 homolog C (C. elegans) |
Gene Aliases: |
B130051O18Rik, Unc5h3, rcm |
Location: |
Chr.3:141465564-141834924 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Unc5c | NM_001293561.1 | 13 | 2232 | NP_001280490.1 |
NM_009472.4 | 12 | 2175 | NP_033498.1 | |
AK031655.1 | 13 | 2199 | BAC27495.1 | |
AK138987.1 | BAE23847.1 | |||
AK141181.1 | 13 | 2198 | BAE24575.1 | |
AK142411.1 | 13 | 2198 | BAE25058.1 | |
AK142974.1 | 13 | 2126 | BAE25236.1 | |
AK164239.1 | 12 | 1976 | BAE37698.1 | |
BC115772.1 | 13 | 2222 | ||
U72634.1 | 12 | 2175 | AAB54103.1 |
More Information
Set Membership: |
Intragenic Exonic |