Assay Details
Target Gene Details
Entrez Gene ID: | 80890 |
Gene Name: | tripartite motif-containing 2 |
Gene Aliases: |
mKIAA0517, narf |
Location: |
Chr.3:84160439-84305832 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Trim2 | NM_001271725.1 | NP_001258654.1 | ||
NM_001271726.1 | NP_001258655.1 | |||
NM_001271727.1 | NP_001258656.1 | |||
NM_001271728.1 | NP_001258657.1 | |||
NM_030706.3 | NP_109631.2 | |||
AB043550.1 | BAB17634.1 | |||
AF220017.1 | AAG53471.1 | |||
AK045410.1 | BAC32350.1 | |||
AK079415.1 | BAC37640.1 | |||
AK141419.1 | BAE24679.1 | |||
AK147275.1 | BAE27812.1 | |||
AK147395.1 | BAE27883.1 | |||
AK164350.1 | BAE37753.1 | |||
AK220343.1 | BAD90242.1 | |||
BC058961.1 | AAH58961.1 |
More Information
Set Membership: |
Intragenic Non-exonic |