Assay Details
Target Gene Details
Entrez Gene ID: | 80877 |
Gene Name: | LPS-responsive beige-like anchor |
Gene Aliases: |
C80285, D3Ertd775e, Lab300, Lba |
Location: |
Chr.3:86223568-86782705 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 22 - Intron 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lrba | NM_001077687.1 | NP_001071155.1 | ||
NM_001077688.1 | NP_001071156.1 | |||
NM_030695.2 | NP_109620.2 | |||
AF187731.1 | AAG14003.1 | |||
AF188506.1 | AAG15400.1 | |||
AF188507.1 | AAG15401.1 |