Assay Details
Target Gene Details
Entrez Gene ID: | 665113 |
Gene Name: | TRAF2 and NCK interacting kinase |
Gene Aliases: |
1500031A17Rik, 4831440I19Rik, AI451411, C530008O15Rik, C630040K21Rik |
Location: |
Chr.3:28263214-28670585 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 23 - Intron 23 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tnik | NM_001163007.1 | NP_001156479.1 | ||
NM_001163008.1 | NP_001156480.1 | |||
NM_001163009.1 | NP_001156481.1 | |||
NM_026910.1 | NP_081186.1 | |||
AK088459.1 | BAC40365.1 | |||
AK122306.1 | BAC65588.2 | |||
BC137799.1 | ||||
BC137800.1 | ||||
BC151003.1 | ||||
BC158011.1 | ||||
BC158059.1 | ||||
CD348964.1 |
More Information
Set Membership: |
Intragenic Non-exonic |