Assay Details
Target Gene Details
Entrez Gene ID: | 242083 |
Gene Name: | protein phosphatase 1 (formerly 2C)-like |
Gene Aliases: |
3222401G13, 5930404J21Rik, AI481720, AW045850, PP2C, PP2Cepsilon, Pp2ce, Ppp2ce, mKIAA4175 |
Location: |
Chr.3:69316918-69560800 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ppm1l | NM_178726.3 | NP_848841.2 | ||
AF117832.1 | AAD17235.1 | |||
AK028275.1 | BAC25853.1 | |||
AK032529.1 | BAC27913.1 | |||
AK035912.1 | BAC29241.1 | |||
AK045724.1 | BAC32472.1 | |||
AK131646.1 | BAE20738.1 | |||
AK142789.1 | ||||
AK147389.1 | ||||
AK147876.1 | BAE28196.1 | |||
AK165160.1 | ||||
AK190317.1 | ||||
AK220523.1 | BAD90308.1 | |||
AY184801.1 | AAO43055.1 | |||
BC096031.1 | AAH96031.1 |
More Information
Set Membership: |
Intragenic Non-exonic |