Assay Details
Target Gene Details
Entrez Gene ID: | 229228 |
Gene Name: | nudix (nucleoside diphosphate linked moiety X)-type motif 6 |
Gene Aliases: |
Asfgf2b |
Location: |
Chr.3:37404982-37420186 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nudt6 | NM_001291044.1 | NP_001277973.1 | ||
NM_153561.3 | NP_705789.1 | |||
AF453427.1 | AAN76847.1 | |||
AF453428.1 | AAN76848.1 | |||
AK031663.1 | 1 | 4305 | ||
AK145299.1 | BAE26354.1 | |||
BC027267.1 | AAH27267.1 | |||
BC146000.1 |
More Information
Set Membership: |
Intragenic Non-exonic |