Assay Details
Target Gene Details
Entrez Gene ID: | 66046 |
Gene Name: | NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 5 |
Gene Aliases: |
0610007D05Rik, AU015782, SGDH |
Location: |
Chr.3:32737063-32751560 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ndufb5 | NM_025316.2 | NP_079592.2 | ||
AK002279.1 | ||||
AK004372.1 | BAB23279.1 | |||
AK005521.1 | ||||
AK013223.1 | BAB28724.1 | |||
AK018562.1 | BAB31277.1 | |||
AK088061.1 | BAC40123.1 | |||
BC025155.1 | AAH25155.1 |
More Information
Set Membership: |
Intragenic Non-exonic |