Assay Details
Target Gene Details
Entrez Gene ID: | 66868 |
Gene Name: | major facilitator superfamily domain containing 1 |
Gene Aliases: |
1200003O06Rik |
Location: |
Chr.3:67582768-67604231 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mfsd1 | NM_025813.3 | NP_080089.1 | ||
AK004586.1 | BAB23391.1 | |||
AK032688.1 | ||||
AK075859.1 | BAC36010.1 | |||
AK135235.1 | BAE22467.1 | |||
AK136108.1 | BAE22824.1 | |||
AK152444.1 | BAE31224.1 | |||
AK159554.1 | BAE35179.1 | |||
AK166645.1 | ||||
BC024891.1 | AAH24891.1 |