Assay Details
Target Gene Details
Entrez Gene ID: | 83398 |
Gene Name: | N-deacetylase/N-sulfotransferase (heparan glucosaminyl) 3 |
Gene Aliases: |
4921531K01Rik, 4930511P15Rik, N-HSST 3, NDST-3 |
Location: |
Chr.3:123526166-123690852 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ndst3 | NM_001293682.1 | 8 | 1076 | NP_001280611.1 |
NM_031186.3 | 9 | 2189 | NP_112463.2 | |
NR_121616.1 | 10 | 2293 | ||
AF221095.1 | 9 | 2188 | AAG34793.1 | |
AK014990.1 | 7 | 1229 | ||
AK015768.1 | 8 | 1076 | BAB29967.1 | |
AK141945.1 | 10 | 2294 | BAE24894.1 | |
BC079622.1 | 9 | 2060 | AAH79622.1 | |
BC112404.1 | 9 | 2101 |
More Information
Set Membership: |
Intragenic Exonic |