Assay Details
Target Gene Details
Entrez Gene ID: | 20416 |
Gene Name: | src homology 2 domain-containing transforming protein C1 |
Gene Aliases: |
Shc, ShcA, p66, p66shc |
Location: |
Chr.3:89418481-89430029 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Shc1 | NM_001113331.2 | NP_001106802.1 | ||
NM_011368.5 | NP_035498.2 | |||
AK036980.1 | ||||
AK049357.1 | BAC33706.1 | |||
AK155158.1 | BAE33083.1 | |||
AK195313.1 | ||||
AK198488.1 | ||||
AK203451.1 | ||||
BC036172.1 | AAH36172.1 | |||
BY341731.1 | ||||
U15784.1 | AAC52146.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |