Assay Details
Target Gene Details
Entrez Gene ID: | 70099 |
Gene Name: | structural maintenance of chromosomes 4 |
Gene Aliases: |
2500002A22Rik, C79747, SMC-4, Smc4l1 |
Location: |
Chr.3:69004972-69034623 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Smc4 | NM_133786.3 | 16 | 2514 | NP_598547.1 |
AJ534940.2 | 15 | 2390 | CAD59183.1 | |
AK048650.1 | 11 | 1727 | ||
BC061481.1 | 16 | 2489 | AAH61481.1 | |
BC062939.1 | 16 | 2514 | AAH62939.1 |