Assay Details
Target Gene Details
Entrez Gene ID: | 17829 |
Gene Name: | mucin 1, transmembrane |
Gene Aliases: |
CD227, EMA, Muc-1 |
Location: |
Chr.3:89229056-89233381 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Muc1 | NM_013605.2 | 6 | 1772 | NP_038633.1 |
AW913641.1 | 2 | 51 | ||
BC005441.1 | 6 | 1743 | AAH05441.1 | |
BG079301.2 | 5 | 401 | ||
M84683.1 | 6 | 1722 | AAA39756.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)