Assay Details
Target Gene Details
Entrez Gene ID: | 18583 |
Gene Name: | phosphodiesterase 7A |
Gene Aliases: |
AU015378, AW047537 |
Location: |
Chr.3:19223108-19311374 on Build GRCm38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pde7a | NM_001122759.2 | NP_001116231.1 | ||
NM_008802.3 | NP_032828.2 | |||
AK036517.1 | ||||
AK154521.1 | BAE32648.1 | |||
AK168957.1 | BAE40763.1 | |||
AY007702.1 | AAG16295.1 | |||
BC062909.1 | AAH62909.1 | |||
CV558025.1 | ||||
U68171.1 | AAB08479.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |