Assay Details
Target Gene Details
Entrez Gene ID: | 114249 |
Gene Name: | nephronectin |
Gene Aliases: |
1110009H02Rik, AA682063, AI314031, Nctn, POEM |
Location: |
Chr.3:132881745-132950291 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Npnt | NM_001029836.2 | NP_001025007.1 | ||
NM_001287101.1 | NP_001274030.1 | |||
NM_001287102.1 | NP_001274031.1 | |||
NM_001287103.1 | NP_001274032.1 | |||
NM_033525.3 | NP_277060.2 | |||
AB059656.1 | BAB69692.1 | |||
AF397007.1 | AAK84391.1 | |||
AF397008.1 | AAK84392.1 | |||
AI527763.1 | ||||
AK050484.1 | BAC34283.1 | |||
AK158764.1 | BAE34649.1 | |||
AK199460.1 | ||||
AK201444.1 | ||||
AY035898.1 | AAK96010.1 | |||
AY035899.1 | AAK96011.1 | |||
BC068308.1 | AAH68308.1 | |||
CK387038.1 |