Assay Details
Target Gene Details
Entrez Gene ID: | 66350 |
Gene Name: | phospholipase A2, group XIIA |
Gene Aliases: |
2310004B05Rik, GXII, GXII-1, GXII-1-PLA2, Pla2g12, Rossy, mGXII-1 |
Location: |
Chr.3:129878606-129895825 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pla2g12a | NM_001286948.1 | NP_001273877.1 | ||
NM_023196.4 | NP_075685.2 | |||
NM_183423.3 | 1 | 53 | NP_904359.1 | |
NR_104611.1 | ||||
AK003183.1 | ||||
AK009133.1 | BAB26094.1 | |||
AK010011.1 | BAB26641.1 | |||
AK010174.1 | BAB26747.1 | |||
AK012414.1 | BAC25368.1 | |||
AK163693.1 | BAE37460.1 | |||
AK178670.1 | ||||
AY007381.1 | AAG23336.1 | |||
AY007382.1 | 1 | 53 | AAG23337.1 | |
BC026812.1 | AAH26812.1 | |||
BC051117.1 | AAH51117.1 |
More Information
Set Membership: |
Intragenic Exonic |