Assay Details
Target Gene Details
Entrez Gene ID: | 320478 |
Gene Name: | SOX2 overlapping transcript (non-protein coding) |
Gene Aliases: |
B230215L15Rik |
Location: |
Chr.3:34560381-34677993 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sox2ot | NR_015580.2 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)