Assay Details
Target Gene Details
Entrez Gene ID: | 229317 |
Gene Name: | eukaryotic translation initiation factor 2A |
Gene Aliases: |
D030048D22, D3Ertd194e |
Location: |
Chr.3:58525821-58557501 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eif2a | NM_001005509.2 | 6 | 666 | NP_001005509.1 |
AK050971.1 | 6 | 383 | ||
AK078519.1 | 6 | 445 | BAC37320.1 | |
AK145275.1 | 6 | 622 | ||
AK161904.1 | 6 | 447 | ||
BC030447.1 | 5 | 335 | ||
BC082557.1 | 6 | 471 | AAH82557.1 |
More Information
Set Membership: |
Intragenic Exonic |