Assay Details
Target Gene Details
Entrez Gene ID: | 20585 |
Gene Name: | helicase-like transcription factor |
Gene Aliases: |
AF010600, BC057116, P113, Smarca3, Snf2l3 |
Location: |
Chr.3:20057811-20118491 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hltf | NM_009210.3 | NP_033236.2 | ||
NR_105046.1 | ||||
NR_105047.1 | ||||
AB221636.1 | ||||
AF010600.1 | AAB63915.1 | |||
AF165911.1 | ||||
AK141235.1 | BAE24607.1 | |||
AK149897.1 | ||||
AK166911.1 | ||||
BC057116.1 | AAH57116.1 | |||
BC059240.1 | AAH59240.1 |