Assay Details
Target Gene Details
Entrez Gene ID: | 229672 |
Gene Name: | BCLl2-like 15 |
Gene Aliases: |
Bfk, Gm566 |
Location: |
Chr.3:103831039-103854618 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bcl2l15 | NM_001142959.1 | NP_001136431.1 | ||
NM_001142960.1 | NP_001136432.1 | |||
AF014453.1 | ||||
BC027668.1 | AAH27668.1 | |||
BC117780.1 | ||||
BC117781.1 | ||||
BC128505.1 | ||||
BC128506.1 |