Assay Details
Target Gene Details
Entrez Gene ID: | 13645 |
Gene Name: | epidermal growth factor |
Gene Aliases: |
AI790464 |
Location: |
Chr.3:129677569-129755322 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Egf | NM_010113.3 | NP_034243.2 | ||
AK085707.1 | ||||
AK088408.1 | ||||
AK136439.1 | BAE22978.1 | |||
AK142666.1 | ||||
BC017681.1 | AAH17681.1 | |||
BC060741.1 | AAH60741.1 | |||
BC092277.1 | AAH92277.1 | |||
J00380.1 | AAA37539.1 | |||
V00741.1 | CAA24115.1 |
More Information
Set Membership: |
Intragenic Non-exonic |