Assay Details
Target Gene Details
Entrez Gene ID: | 13645 |
Gene Name: | epidermal growth factor |
Gene Aliases: |
AI790464 |
Location: |
Chr.3:129677569-129755322 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Egf | NM_010113.3 | 13 | 2282 | NP_034243.2 |
AK085707.1 | 13 | 1849 | ||
AK088408.1 | 8 | 869 | ||
AK136439.1 | 8 | 875 | BAE22978.1 | |
AK142666.1 | 13 | 1978 | ||
AK157778.1 | 8 | 874 | ||
BC017681.1 | 14 | 2219 | AAH17681.1 | |
BC060741.1 | 13 | 2267 | AAH60741.1 | |
BC092277.1 | 13 | 2267 | AAH92277.1 | |
J00380.1 | 13 | 2277 | AAA37539.1 | |
V00741.1 | 13 | 2233 | CAA24115.1 |
More Information
Set Membership: |
Intragenic Exonic |