Assay Details
Target Gene Details
Entrez Gene ID: | 329731 |
Gene Name: | family with sequence similarity 19, member A3 |
Gene Aliases: |
7530404M11Rik, TAFA3, Tafa-3 |
Location: |
Chr.3:104767405-104777547 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fam19a3 | NM_183224.3 | NP_899047.1 | ||
AK078681.1 | ||||
AY325122.1 | AAP92414.1 | |||
BC118531.1 |
More Information
Set Membership: |
Intragenic Non-exonic |