Assay Details
Target Gene Details
Entrez Gene ID: | 68259 |
Gene Name: | intraflagellar transport 80 |
Gene Aliases: |
4921524P20Rik, Wdr56, mKIAA1374 |
Location: |
Chr.3:68892499-69004653 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ift80 | NM_026641.2 | 8 | 894 | NP_080917.1 |
AK042231.1 | 8 | 819 | BAC31200.1 | |
AK129339.1 | 8 | 822 | BAC98149.1 | |
AK147305.1 | 7 | 4421 | ||
BC013814.1 | 3 | 183 | AAH13814.1 | |
BC034101.1 | 8 | 789 | AAH34101.1 |