Assay Details
Target Gene Details
Entrez Gene ID: | 13601 |
Gene Name: | extracellular matrix protein 1 |
Gene Aliases: |
AI663821, p85 |
Location: |
Chr.3:95734147-95739819 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ecm1 | NM_001252653.1 | NP_001239582.1 | ||
NM_007899.2 | NP_031925.2 | |||
AK153009.1 | BAE31648.1 | |||
AK153471.1 | BAE32022.1 | |||
AK159334.1 | BAE34999.1 | |||
AK169939.1 | BAE41471.1 | |||
BC138693.1 | ||||
BC145381.1 | ||||
L33416.1 | AAA37535.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |