Assay Details
Target Gene Details
Entrez Gene ID: | 11770 |
Gene Name: | fatty acid binding protein 4, adipocyte |
Gene Aliases: |
422/aP2, ALBP/Ap2, Ap2, Lbpl |
Location: |
Chr.3:10204342-10208597 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fabp4 | NM_024406.2 | NP_077717.1 | ||
AK003143.1 | BAB22601.1 | |||
AK088535.1 | BAC40410.1 | |||
BC054426.1 | AAH54426.1 | |||
BY081693.1 | ||||
CT010390.1 | CAJ18597.1 | |||
K02109.1 | AAA39416.1 |