Assay Details
Target Gene Details
Entrez Gene ID: | 14866 |
Gene Name: | glutathione S-transferase, mu 5 |
Gene Aliases: |
- |
Location: |
Chr.3:107895854-107898685 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gstm5 | NM_010360.2 | NP_034490.1 | ||
AK002750.1 | BAB22327.1 | |||
AK019317.1 | BAB31661.1 | |||
AK028098.1 | BAC25746.1 | |||
BC008206.1 | AAH08206.1 | |||
U24428.1 | AAB04096.1 |
More Information
Set Membership: |
Intragenic Non-exonic |