Assay Details
Target Gene Details
Entrez Gene ID: | 26377 |
Gene Name: | dual adaptor for phosphotyrosine and 3-phosphoinositides 1 |
Gene Aliases: |
Bam32 |
Location: |
Chr.3:137930998-137981576 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dapp1 | NM_011932.2 | 4 | 556 | NP_036062.1 |
AF163255.1 | 4 | 510 | AAD49698.1 | |
AF186023.1 | 4 | 531 | AAF14579.1 | |
AK076886.1 | 2 | 109 | ||
AK145745.1 | 4 | 522 | BAE26622.1 | |
AK146180.1 | 4 | 525 | BAE26958.1 | |
AK169584.1 | 3 | 421 | BAE41242.1 | |
BC014759.1 | 4 | 552 | AAH14759.1 |