Assay Details
Target Gene Details
Entrez Gene ID: | 12167 |
Gene Name: | bone morphogenetic protein receptor, type 1B |
Gene Aliases: |
AI385617, ALK-6, AV355320, Acvrlk6, Alk6, BMPR-1B, BMPR-IB, CFK-43a, SKR6 |
Location: |
Chr.3:141837136-142169590 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bmpr1b | NM_001277216.1 | NP_001264145.1 | ||
NM_001277217.1 | NP_001264146.1 | |||
NM_001277218.1 | NP_001264147.1 | |||
NM_001277220.1 | NP_001264149.1 | |||
NM_007560.4 | NP_031586.1 | |||
AK086130.1 | BAC39617.1 | |||
AK140158.1 | BAE24260.1 | |||
AK162844.1 | BAE37078.1 | |||
BC065106.1 | AAH65106.1 | |||
BC065143.1 | AAH65143.1 | |||
BC138638.1 | ||||
BC138640.1 | ||||
CV557650.1 | ||||
Z23143.1 | CAA80674.1 |
More Information
Set Membership: |
Intragenic Non-exonic |