Assay Details
Target Gene Details
Entrez Gene ID: | 66926 |
Gene Name: | tRNA methyltransferase 6 |
Gene Aliases: |
3300001M20Rik, AU016412, AW492787, CGI-09, mKIAA1153 |
Location: |
Chr.2:132804207-132816054 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Trmt6 | NM_175113.3 | NP_780322.2 | ||
AK014359.1 | BAC25432.1 | |||
AK028755.1 | BAC26100.1 | |||
AK129300.1 | BAC98110.1 | |||
AK164396.1 | ||||
AK165976.1 | BAE38495.1 | |||
AK167222.1 | BAE39347.1 | |||
BC049083.1 | 5 | 1656 | AAH49083.1 | |
BC052648.1 | AAH52648.1 |
More Information
Set Membership: |
Intragenic Non-exonic |