Assay Details
Target Gene Details
Entrez Gene ID: | 73668 |
Gene Name: | tetratricopeptide repeat domain 21B |
Gene Aliases: |
2410066K11Rik, Thm1, aln, mKIAA1992 |
Location: |
Chr.2:66184327-66256675 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ttc21b | NM_001047604.2 | NP_001041069.1 | ||
NM_001290669.1 | NP_001277598.1 | |||
AK010712.1 | ||||
AK147810.1 | BAE28153.1 | |||
DQ011164.1 |