Assay Details
Target Gene Details
Entrez Gene ID: | 71856 |
Gene Name: | WAP four-disulfide core domain 3 |
Gene Aliases: |
1700015L13Rik, 1700127F16Rik |
Location: |
Chr.2:164731226-164745910 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Wfdc3 | NM_027961.1 | NP_082237.1 | ||
AK007299.1 | ||||
AY974580.1 | ||||
BC116847.1 | ||||
BC116875.1 |
More Information
Set Membership: |
Intragenic Non-exonic |