Assay Details
Target Gene Details
Entrez Gene ID: | 67701 |
Gene Name: | WAP four-disulfide core domain 2 |
Gene Aliases: |
1600023A02Rik, HE4, WAP5 |
Location: |
Chr.2:164562563-164568510 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Wfdc2 | NM_026323.2 | 5 | 579 | NP_080599.1 |
AF334269.1 | 5 | 576 | AAL73189.1 | |
AK003771.1 | 3 | 222 | ||
AK005519.1 | 5 | 580 | BAB24094.1 | |
BC099427.1 | 5 | 570 | AAH99427.1 |
More Information
Set Membership: |
Intragenic Exonic |