Assay Details
Target Gene Details
Entrez Gene ID: | 380601 |
Gene Name: | FAST kinase domains 5 |
Gene Aliases: |
C78212, mKIAA1792 |
Location: |
Chr.2:130613838-130630027 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fastkd5 | NM_001146084.1 | NP_001139556.1 | ||
NM_198176.2 | NP_937819.2 | |||
AK143929.1 | BAE25612.1 | |||
BB610359.1 |
Target Gene Details
Entrez Gene ID: | 140629 |
Gene Name: | U box domain containing 5 |
Gene Aliases: |
1500010O06Rik, C330018L13Rik, Rnf37, UIP5, Ubce7ip5, Ube7ip5 |
Location: |
Chr.2:130589996-130630038 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ubox5 | NM_001255993.1 | NP_001242922.1 | ||
NM_001255994.1 | NP_001242923.1 | |||
NM_080562.5 | NP_542129.2 | |||
AF360997.1 | AAK51467.1 | |||
AK005195.1 | ||||
AK034114.1 | BAC28590.1 | |||
AK158052.1 | BAE34338.1 | |||
AK170024.1 | ||||
AK170100.1 | ||||
AK171070.1 |