Assay Details
Target Gene Details
Entrez Gene ID: | 277353 |
Gene Name: | transcription factor-like 5 (basic helix-loop-helix) |
Gene Aliases: |
AV260129, Figlb |
Location: |
Chr.2:180621956-180642691 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tcfl5 | NM_178254.3 | 4 | 1266 | NP_839985.2 |
AK132721.1 | 4 | 926 | BAE21320.1 | |
AY234363.1 | 4 | 1060 | AAO75014.1 | |
BC108399.1 | 4 | 1266 | AAI08400.1 |
More Information
Set Membership: |
Intragenic Exonic |