Assay Details
Target Gene Details
Entrez Gene ID: | 229211 |
Gene Name: | acyl-Coenzyme A dehydrogenase family, member 9 |
Gene Aliases: |
2600017P15Rik, 4732402K02, C630012L17Rik, NPD002 |
Location: |
Chr.3:36065984-36092857 on Build GRCm38 |
Assay Gene Location: | Within Intron 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Acad9 | NM_172678.3 | NP_766266.3 | ||
AK012773.1 | BAB28460.2 | |||
AK028571.1 | ||||
AK031820.1 | BAC27565.1 | |||
AK075984.1 | BAC36096.1 | |||
AK088409.1 | BAC40337.1 | |||
AK145423.1 | BAE26429.1 | |||
AK181172.1 | ||||
BC032213.1 | AAH32213.1 | |||
BC033277.1 | AAH33277.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |