Assay Details
Target Gene Details
Entrez Gene ID: | 68369 |
Gene Name: | RIKEN cDNA 0610031O16 gene |
Gene Aliases: |
- |
Location: |
Chr.3:138210717-138239663 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
0610031O16Rik | NR_045760.1 | |||
AK002733.1 |
Target Gene Details
Entrez Gene ID: | 11529 |
Gene Name: | alcohol dehydrogenase 7 (class IV), mu or sigma polypeptide |
Gene Aliases: |
AI325182, Adh-3, Adh-3e, Adh-3t, Adh3, Adh3-e, Adh3-t, Adh4, Adt-1 |
Location: |
Chr.3:138198053-138232042 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Adh7 | NM_009626.4 | NP_033756.2 | ||
AF331802.1 | AAM22808.1 | |||
AF331803.1 | AAM22809.1 | |||
AK009588.2 | BAE43215.1 | |||
AK009600.1 | BAB26385.1 | |||
BC047267.1 | AAH47267.1 | |||
U20257.1 | AAA76735.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |